Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ LiChroCART™ LiChrospher™ RP-18 C18 ADS Mixed Mode HPLC Column, 25 μm, 4 mm x 25 mm
Click to view available options
Length (Metric):
25 mm
Description
LiChrospher™ ADS allows the fully automated, LC integrated sample preparation of hydrophobic, low molecular weight analytes from untreated samples like serum, milk, fermentation broth of food homogenates. The enrichment is based on two chromatographic processes: reversed phase and size exclusion chromatography since LiChrospher™ ADS sorbents are spherical silica gel particles (25μm) with DIOL modification on the exterior surface and alkyl modifications on the surface of the interior pores (ID: 6nm). Depending on the hydrophobicity of the analyte three types of precolumns are available: C-4, C-8 and C-18.

Specifications
Specifications
Column Format | Cartridge |
Diameter (Metric) | 4 mm |
Length (Metric) | 25 mm |
Particle Size | 25 μm |
pH | 2 to 7.5 |
Pore Size | 60 Å |
Quantity | 3/Pk. |
USP Type | L1 |
Stationary Phase | C18 ADS |
Packing Material | Silica |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction