Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eNOS Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-5068092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300500 100 μL
NB300500SS 0.02 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300500 Supplier Novus Biologicals Supplier No. NB300500
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 16 publications

eNOS Polyclonal specifically detects eNOS in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Drosophila, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen eNOS
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:50-1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P29474
Gene Alias Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII
Gene Symbols NOS3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues S(1179) LQERQLRGAVPWAFD(1194) of human eNOS.
Molecular Weight of Antigen 133 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4846
Test Specificity This does not detect human inducible NOS or rat brain NOS.
Target Species Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Drosophila, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.