Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EnzChek™ Phosphatase Assay Kit

Catalog No. e12020
Encompass_Preferred
Click to view available options
Quantity:
1000 Assays

Continuous assay of prostatic acid phosphatase and protein phosphatase 1

Using the EnzChek Phosphatase Assay Kit continuous monitoring of phosphatase activity, even at neutral or acidic pH, can be achieved. The kit contains Molecular Probes (proprietary) 6,8-difluoro-4-methylumbelliferyl phosphate (DiFMUP) substrate. DiFMUP is about 100 times more sensitive than 4-methylumbelliferyl phosphate (MUP) for the detection of prostatic acid phosphatase at pH 5.5. The kit is perfect for the continuous assay of prostatic acid phosphatase, protein phosphatase 1 or almost any other phosphatase that can be assayed with nonprotein-based substrates such as MUP or 4-nitrophenyl phosphate (PNPP). Each kit contains a sufficient amount of substrate for ∽1000 assays, using a reaction volume of 100μL per assay. Fluorescence (excitation/emission maxima approx. 358/455nm) can be measured in a fluorescence microplate reader or a standard fluorometer.

Cell Analysis, Cellular Imaging, Drug Discovery and Development, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Phosphatase Assay
For Use With (Equipment) Microplate Reader
Product Type Phosphatase Assay
Dye Type DiFMU
Emission 360/460
Product Line EnzChek
Quantity 1000 Assays
Shipping Condition Room Temperature
Substrate DiFMUP
Substrate Properties Chemical Substrate
Substrate Type Phosphatase Substrate
Target Enzyme Phosphatases
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.