Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

58K Golgi Protein Antibody (58K-9), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$192.94 - $493.10

Specifications

Antigen 58K Golgi Protein
Clone 58K-9
Dilution Western Blot 1:2000 - 1:5000, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 24505439)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600412SS
SDP
View Documents
Novus Biologicals
NB600412SS
0.025 mL
Each for $192.94
Only null left
Add to Cart
 
NB600412
SDP
View Documents
Novus Biologicals
NB600412
0.1 mL
Each for $493.10
Only null left
Add to Cart
 
Description

Description

58K Golgi Protein Monoclonal specifically detects 58K Golgi Protein in Human, Mouse, Rat, Bovine, Canine, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

58K Golgi Protein
Western Blot 1:2000 - 1:5000, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 24505439)
Monoclonal
Ascites
RUO
Human, Mouse, Rat, Bovine, Canine, Chicken
O95954, Q91XD4, O88618
10841
58K Golgi Protein purified from rat liver [UniProt# O88618]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
58 kDa
58K-9
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism, Signal Transduction
Ascites with 0.1% Sodium Azide
formimidoyltransferase cyclodeaminase, formimidoyltransferase-cyclodeaminase, formiminotransferase cyclodeaminase, Formiminotransferase-cyclodeaminase, human formiminotransferase cyclodeaminase, EC 4.3.1.410formiminotransferase-cyclodeaminase, LCHC1
FTCD
IgG1
Unpurified
Rat's 58K Golgi Protein sequence is 92% similar with Chinese hamster (Cricetulus griseus).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.