Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epithelial Specific Antigen/Ep-CAM Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-1, Mouse Monoclonal Antibody.

$543.00

Specifications

Antigen Epithelial Specific Antigen/Ep-CAM Ab-1
Clone VU-1D9
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Mouse
View More Specs

Products 10
Catalog Number Mfr. No. Quantity Conjugate Form Concentration Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Form Concentration Price Quantity & Availability  
MS144P1
View Documents
Epredia
MS144P1
500 μL Unconjugated Concentrated 200μg/mL
Each for $543.00
Only null left
Add to Cart
 
MS144B0
View Documents
Epredia
MS144B0
100 μL Biotin Concentrated 200μg/mL N/A N/A N/A
MS144B1
View Documents
Epredia
MS144B1
500 μL Biotin Concentrated 200μg/mL N/A N/A N/A
MS144B
View Documents
Epredia
MS144B
1 mL Biotin Concentrated 200μg/mL N/A N/A N/A
MS144P0
View Documents
Epredia
MS144P0
100 μL Unconjugated Concentrated 200μg/mL N/A N/A N/A
MS144P1ABX
View Documents
Epredia
MS144P1ABX
100 μL Unconjugated Concentrated 1mg/mL N/A N/A N/A
MS144PABX
View Documents
Epredia
MS144PABX
200 μL Unconjugated Concentrated 1mg/mL N/A N/A N/A
MS144P
View Documents
Epredia
MS144P
1 mL Unconjugated Concentrated 200μg/mL N/A N/A N/A
MS144R1
View Documents
Epredia
MS144R1
1 mL Unconjugated Prediluted N/A N/A N/A
MS144R7
View Documents
Epredia
MS144R7
7 mL Unconjugated Prediluted N/A N/A N/A
Description

Description

EGP40 is a 40 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. It reportedly distinguishes adenocarcinomas from pleural mesotheliomas.

Host Species: Mouse

Clone: VU-1D9

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Small cell lung carcinoma cells

Molecular Weight: 40-43 kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Epithelial Specific Antigen/Ep-CAM Ab-1
Immunohistochemistry (Paraffin)
Mouse
Cancer and Tumor Biology
Small cell lung carcinoma cells
Primary
VU-1D9
Monoclonal
IVD
Human
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.