Learn More
Eppendorf™ Trade-in Xplorer™ Electronic Pipetters, Multichannel
Shop All Eppendorf North America Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
15 to 300 μL
5 to 100 μL
50 to 1200 μL
Number of Channels:
8
12
Color:
Gray
Green
Orange
Yellow
Description
Pipetters are based on the proven Eppendorf PhysioCare™ concept. The features set new standards for simplicity, precision and reproducibility, which mean no more delays due to complicated programming or inflexible processes. Precisely adjustable parameters, maximally reproducible results, fatigue-free work and full control over the pipetting processes are achieved.
- Intuitive user interface
- Pipetting modes: autodispensing, dispensing, pipetting, mixing, manual pipetting
- Universal nose cone fits most pipette tips
- Highly reproducible results
- Spring engaged nose cone for enhanced tip attachment
- Light weight design offers unsurpassed ergonomics
Specifications
Specifications
Color | Gray |
Number of Channels | 8 |
Product Line | Trade-in Xplorer™ |
Type | Electronic Pipette |
Volume (Metric) | 0.5 to 10 μL |
Finger Style | Ergonomic |