Learn More
Eppendorf™ Trade-in Xplorer™ Electronic Pipetter Available on GSA/VA Contract for Federal Government customers only.
Specially designed for high professional standards to provide optimal support
Manufacturer:  Eppendorf™ 486100164
 View more versions of this product
Catalog No. 14-286-543
Add to cart
Description
Description
Pipetters are based on the proven Eppendorf PhysioCare™ concept. The features set new standards for simplicity, precision and reproducibility, which mean no more delays due to complicated programming or inflexible processes. Precisely adjustable parameters, maximally reproducible results, fatigue-free work and full control over the pipetting processes are achieved.
  • Intuitive user interface
  • Pipetting modes: autodispensing, dispensing, pipetting, mixing, manual pipetting
  • Universal nose cone fits most pipette tips
  • Highly reproducible results
  • Spring engaged nose cone for enhanced tip attachment
  • Light weight design offers unsurpassed ergonomics
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Ergonomic
Green
8
50 to 1200μL