Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ER Membrane Protein Complex Subunit 10 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP275482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275482 Supplier Novus Biologicals Supplier No. NBP275482

Rabbit Polyclonal Antibody

ER Membrane Protein Complex Subunit 10 Polyclonal specifically detects ER Membrane Protein Complex Subunit 10 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ER Membrane Protein Complex Subunit 10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
Gene Alias C19orf63, Chromosome 19 Open Reading Frame 63, EMC10, Hematopoietic Signal Peptide-Containing Membrane Domain-Containing 1, Hematopoietic Signal Peptide-Containing Membrane Domain-Containing Protein 1, Hematopoietic Signal Peptide-Containing Secreted 1, HSM1, HSS1, INM02, UPF0510 Protein INM02
Gene Symbols EMC10
Host Species Rabbit
Immunogen Recombinant protein within mouse ER Membrane Protein Complex Subunit 10 aa 1-258/ 258. (SwissProt: Q5UCC4 Human; SwissProt: Q3TAS6 Mouse)
Molecular Weight of Antigen 27 kDa
Purification Method Immunogen affinity purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 284361.0
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.