Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ERK1 Inhibitor Peptide Set
SDP

For use in research applications

$572.50 - $1244.00

Specifications

Host Species Human, Mouse, Rat, Hamster, Rabbit, Xenopus
Components ERK Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKGMPKKKPTPIQLN (ERK inhibitor sequence: GMPKKKPTPIQLN). Molecular weight: 3795. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
For Use With (Application) Inhibition of Erk activation
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Product Type ERK1 Inhibitor Peptide Set
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2293335
SDP
View Documents
Novus Biologicals™
NBP2293335MG
5 mg
Each for $1,244.00
Only null left
Add to Cart
 
NBP229333
SDP
View Documents
Novus Biologicals™
NBP229333
2 mg
Each for $572.50
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

Human, Mouse, Rat, Hamster, Rabbit, Xenopus
Inhibition of Erk activation
ERK1 Inhibitor Peptide Set
ERK1
ERK Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKGMPKKKPTPIQLN (ERK inhibitor sequence: GMPKKKPTPIQLN). Molecular weight: 3795. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
3795
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.