Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ether, Anhydrous, min. 99.0%, BAKER ANALYZED™ ACS Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Description
- Bulk Pharmaceutical Chemical
Specifications
Specifications
| Density | 0.7079g/mL |
| Flash Point | −45°C (− 113°C) |
| Packaging | Aluminum Bottle |
| Quantity | 1 L |
| Residue after Evaporation | max. 0.001% |
| Specific Gravity | 1L = 0.71kg |
| Percent Purity | ≥99.0% |
| Grade | Meets USP requirements |
| Physical Form | Liquid |
| Chemical Name or Material | Anhydrous, Contains BHT as a Preservative |
Safety and Handling
| Recommended Storage | Keep in partly filled, tightly closed, light resistant containers, at a temperature not exceeding 30°C (86°F), remote from fire |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction