Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Preserved with 2% Alcohol, 0.5% max Water and 7ppm min BHT, U.S.P., J.T. Baker™

Catalog No. p-5391128 Shop All Avantor Products
Click to view available options
Quantity:
1 L
Packaging:
Aluminum Bottle

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Flash Point −45.00°C
Packaging Aluminum Bottle
Quantity 1 L
Specific Gravity 1L = 0.71kg
Formula Weight 74.12
Grade USP
Chemical Name or Material Ether, Preserved with 2% Alcohol, 0.5% max Water and 7ppm min BHT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.