Learn More
MSA™ Evotech™ Standard Full Body Harnesses
Catalog No. p-4512025 Shop All MSA Products
Change view
Click to view available options
Buckle Style (Leg):
Quick-Connect
Tongue-buckle leg strap
Certifications/Compliance:
ANSI Z359.1, A10.32, CSA 259.10-06, OSHA
CSA Z259.10-06; Meets OSHA and ANSI Z359.1
D-Ring Location:
Back
Back, Side
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style (Leg) Certifications/Compliance D-Ring Location
17-200-116 Tongue-buckle leg strap ANSI Z359.1, A10.32, CSA 259.10-06, OSHA Back
17-200-117 Tongue-buckle leg strap ANSI Z359.1, A10.32, CSA 259.10-06, OSHA Back, Side
17-200-118 Quick-Connect CSA Z259.10-06; Meets OSHA and ANSI Z359.1 Back
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-200-116 Supplier MSA™ Supplier No. 10105931
Exceptional comfort, ease of use, durability and user safety
  • No binding edge on shoulder padding to prevent neck chafing
  • Breathable padding with moisture-wicking material keeps users cooler
  • Leg strap design keeps leg straps in place to increase mobility and comfort
  • Variable width sub-pelvic webbing provides 50% more surface area for better weight distribution
  • Dual-color webbing (red pattern outside, black inside) simplifies harness donning
  • Web management system conveniently stores excess webbing
  • Positionable lanyard clips
  • RFID enabled to allow easy tracking and inspection
Certifications
CSA Z259.10-06
Compliance
Meets OSHA and ANSI Z359.1
Specifications
Style Subpelvic full body
Product Type Subpelvic Full-body Harness
Includes Manual
D-Ring Location Back
Buckle Style (Chest) Quick-Connect
For Use With (Application) Construction
Webbing Material Polyester
Capacity (English) 400 lb.
Certifications/Compliance ANSI Z359.1, A10.32, CSA 259.10-06, OSHA
Buckle Style (Leg) Tongue-buckle leg strap
Size Standard
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.