Learn More
Invitrogen™ EXPRESS qPCR Supermix, universal
EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for probe-based qPCR assays.
$948.65 - $4316.65
Specifications
Detection Method | Primer-probe |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Reaction Speed | Fast or Standard |
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
11-785-200
![]() |
Invitrogen™
11785200 |
200 Reactions |
Each
for $948.65
Save 3% Reg : |
|
|||||
11-785-01K
![]() |
Invitrogen™
1178501K |
2500 Reactions |
Each
for $4,316.65
Save 3% Reg : |
|
|||||
Description
- EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for probe-based qPCR assays
- This kit comes with a separate tube of ROX reference dye
- EXPRESS qPCR SuperMix kits with premixed ROX reference dye are also available
This kit provides
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
Primer-probe | |
High | |
Fast or Standard | |
Gene Expression |
Tube | |
qPCR | |
Express qPCR Supermix | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.