Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZFIT/ZNF71 Antibody, Novus Biologicals™
SDP

Catalog No. NBP180358 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP180358 100 μL
NBP18035820 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP180358 Supplier Novus Biologicals Supplier No. NBP180358
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EZFIT/ZNF71 Polyclonal specifically detects EZFIT/ZNF71 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EZFIT/ZNF71
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_067039
Gene Alias endothelial zinc finger protein induced by tumor necrosis factor alpha, Kruppel-related zinc finger protein, zinc finger protein 71 (Cos26), zinc finger protein 71EZFITCos26
Gene Symbols ZNF71
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF71. Peptide sequence RCGQCGKSFIKNSSLTVHQRIHTGEKPYRCGECGKTFSRNTNLTRHLRIH.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58491
Test Specificity Expected identity based on immunogen sequence: Equine: 100%; Human: 100%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.