Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-box protein 15/FBXO15 Antibody [Janelia Fluor™ 669], Novus Biologicals™
SDP

Catalog No. AF6035JF669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6035JF669 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF6035JF669 Supplier Novus Biologicals Supplier No. AF6035JF669
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

F-box protein 15/FBXO15 Polyclonal antibody specifically detects F-box protein 15/FBXO15 in Human samples. It is validated for Western Blot

Specifications

Antigen F-box protein 15/FBXO15
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias F-box only protein 15, F-box protein 15, FBX15MGC39671
Host Species Sheep
Immunogen E. coli-derived recombinant human F-box protein 15/FBXO15, aa 298-434, Accession # NP_689889
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201456
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.