Learn More
F4/80 Antibody (Cl:A3-1 (recombinant version)), Allophycocyanin, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | F4/80 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Cl:A3-1 (recombinant version) |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 |
| Gene Symbols | Adgre1 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.