Learn More
Kimberly-Clark™ Professional Faceshields
Kimberly-Clark™ Professional Faceshields are designed to provide on-the-job protection for heat applications, as well as for grinding, flying debris and splashes.
Specifications
Width (English) 15.5 in.
Product Type Faceshield
View More Specs
Products 4
Catalog Number Mfr. No. Length (English) Lens Material Lens Tint Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Lens Material Lens Tint Width (English) Price Quantity & Availability  
19-169-154
Encompass
View Documents
Kimberly-Clark™ Professional
26262
8 in. Polycarbonate Clear, IR/UV 3.0 15.5 in. N/A N/A
Please call customer service for current pricing. 1-800-766-7000
19-169-155
Encompass
View Documents
Kimberly-Clark™ Professional
29080
8 in. Polycarbonate Clear, IR Filter Shade 5.0 15.5 in. N/A N/A
Please call customer service for current pricing. 1-800-766-7000
19-169-100
Encompass
View Documents
Kimberly-Clark™ Professional
29100
8 in. Polycarbonate Green 15.5 in. Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area. N/A
19-169-153
View Documents
Kimberly-Clark™ Professional
29079
9 in. Acetate Clear 15.5 in. Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area. N/A
Description
Description
  • Made of durable polycarbonate
  • May be reused (depending on the task)
  • D3 rating for splash protection and the polycarbonate material provides 99.9% UVA/UVB/UVC protection
  • Compatible with the Jackson Safety Hard Hat Interchange System
  • Snaps into place on compatible head gear and hard hats – no special tools required
  • When used with any Jackson Safety Brand Headgear, meets the performance standards of ANSI Z87.1+ for high-impact protection and is CSA Z94.3 compliant (when used with compatible Jackson Safety headgear)
Specifications
Specifications
15.5 in.
Faceshield
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.