Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM55D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | FAM55D |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17420620
![]() |
Novus Biologicals
NBP17420620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP174206
![]() |
Novus Biologicals
NBP174206 |
100 μL |
Each for $487.50
|
|
|||||
Description
FAM55D Polyclonal specifically detects FAM55D in Mouse samples. It is validated for Western Blot.Specifications
FAM55D | |
Polyclonal | |
Rabbit | |
Q52KP5 | |
54827 | |
Synthetic peptides corresponding to the C terminal of Fam55d. Immunizing peptide sequence NSDVERFSDFHGYTQYLALKDIFQDLNVGVIDAWDMTVAYGINNVHPPED. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
C11orf33, chromosome 11 open reading frame 33, family with sequence similarity 55, member D, FLJ20127, hypothetical protein LOC54827 | |
NXPE4 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title