Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RII/RIII (CD32/CD16) Antibody (190909), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB175205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175205 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175205 Supplier Novus Biologicals Supplier No. FAB1460J
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Fc gamma RII/RIII (CD32/CD16) Monoclonal antibody specifically detects Fc gamma RII/RIII (CD32/CD16) in Mouse samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen Fc gamma RII/RIII (CD32/CD16)
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 190909
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIIB, Thr30-Arg207, Accession # P08101
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2214
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.