Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferritin Heavy Chain Antibody (962610) - Azide and BSA Free, R&D Systems™
SDP

Catalog No. MAB9354SP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9354SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB9354SP Supplier Novus Biologicals Supplier No. MAB9354SP
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ferritin Heavy Chain Monoclonal specifically detects Ferritin Heavy Chain in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Ferritin Heavy Chain
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 962610
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5 ug/ml, Immunocytochemistry 1 ug/ml
Formulation PBS with No Preservative
Gene Alias apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15
Gene Symbols FTH1
Host Species Mouse
Immunogen The immunogen is raised against Ferritin Heavy Chain.
Purification Method Protein A or G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2495
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.