Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR-4, Rabbit anti-Human, Clone: 29.2, Millipore Sigma™
Rabbit Monoclonal Antibody
$192.34 - $498.79
Specifications
Antigen | FGFR-4 |
---|---|
Clone | 29.2 |
Dilution | Flow Cytometry Analysis: A 1:4 dilution from a representative lot detected FGFR-4 in one million LNCaP cells. |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC112725
|
MilliporeSigma
MABC112725UL |
25 μL |
Each for $192.34
|
|
|||||
MABC1127100
|
MilliporeSigma
MABC1127100UL |
100 μL |
Each for $498.79
|
|
|||||
Description
Anti-FGFR-4, clone 29.2, Cat. No. MABC1127, is a rabbit monoclonal antibody that detects Fibroblast growth factor receptor 4 and has been tested for use in Flow Cytometry.
Fibroblast growth factor receptor 4 (UniProt: P22455; also known as EC: 2.7.10.1, FGFR-4, CD334) is encoded by the FGRF4 (also known as JTK2, TKF) gene (Gene ID: 2264) in human. FGFR-4 is a highly conserved, single-pass type I membrane protein with tyrosine kinase activity that serves as a cell surface receptor for fibroblast growth factors and regulates cell proliferation differentiation, and migration. FGFR-4 is expressed in gastrointestinal epithelial cells, pancreas, and gastric and pancreatic cancer cell lines. In the absence of a ligand, it is present as a monomer in an inactive conformation. Upon ligand binding it homodimerizes into an active conformation with autophosphorylation of tyrosine residues. FGFR-4 is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off in the mature form. It contains an extracellular domain (aa 22-369), a transmembrane domain (aa 370-390), and a cytoplasmic domain (aa 391-802). FGFR-4 undergoes N-glycosylation, and when not fully glycosylated it can undergo proteasomal degradation. FGFR-4 signaling is down-regulated by receptor internalization and degradation. MMP14 is reported to promote internalization and degradation of FGFR-4. Mutations that lead to constitutive kinase activation or impair normal FGFR-4 inactivation lead to aberrant signaling leading to tumor growth. (Ref.: Turkington, RC et al. (2014). Cell Death Dis. 5; e1046).Specifications
FGFR-4 | |
Flow Cytometry Analysis: A 1:4 dilution from a representative lot detected FGFR-4 in one million LNCaP cells. | |
Monoclonal | |
Supernatant | |
RUO | |
Human | |
Fibroblast growth factor receptor 4;EC: 2.7.10.1;CD334 | |
FGFR4 | |
IgG | |
Unpurified | |
This rabbit monoclonal antibody detects human fibroblast growth factor receptor 4 (FGFR-4). It targets an epitope with in the extracellular domain. |
29.2 | |
Flow Cytometry | |
Unconjugated | |
Rabbit | |
Signaling | |
Rabbit monoclonal antibody in supernatant without preservatives. | |
NP_001278909 | |
His-tagged recombinant extracellular domain of human fibroblast growth factor receptor 4 (FGFR-4). | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title