Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
fHbp Mouse anti-Bacteria, Clone: JAR 4, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $463.74
Specifications
Antigen | fHbp |
---|---|
Clone | JAR 4 |
Concentration | 1.0 mg/mL |
Applications | ELISA, Flow Cytometry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF267025
|
MilliporeSigma
MABF267025UG |
25 μg |
Each for $182.31
|
|
|||||
MABF2670100
|
MilliporeSigma
MABF2670100UG |
100 μg |
Each for $463.74
|
|
|||||
Description
fHbp Monoclonal antibody specifically detects fHbp in Bacteria samples. It is validated for Activity Assay, ELISA, Flow Cytometry, Western Blotting.Specifications
fHbp | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG2a in PBS without azide | |
IgG2a | |
Protein G purified |
JAR 4 | |
ELISA, Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
Bacteria | |
His-tagged recombinant Factor H-binding protein (fHbp) from variant group 1 expressed in E. coli. | |
Primary | |
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title