Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 506/780 Sample Pack
Click to view available options
Quantity:
1 kit
Description
- Fixable Viability Dyes may be used to label cells from all species
- Supplied as pre-diluted solutions prepared in high-quality, anhydrous DMSO. It should be protected from light and moisture
- Store at −70°C with dessicant
- It may be freeze-thawed up to 20 times
- Allow vial to equilibrate to room temperature before opening
- Can be excited by the violet (405nm) laser line and has a peak emission of 506nm
- For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
- Can be excited by the red (633nm) laser line and has a peak emission of 780nm
- For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
- Staining may be done before or after surface staining
Specifications
Specifications
| Color | Green, Infrared |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Dye |
| Emission | 405/506, 633/780 |
| Product Line | eBioscience |
| Quantity | 1 kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction