Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 450

Click to view available options
Quantity:
100 Tests
500 Tests
Description
- Unlike 7-AAD and propidium iodide, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized, and stained for intracellular antigens without any loss of staining intensity of the dead cells
- Thus, using these dyes allows dead cells to be excluded from analysis when intracellular targets are being studied
- Fixable Viability Dye eFluor™ 450 can be excited by the violet (405 nm) laser line and has a peak excitation of 450 nm that can be detected using a 450/50 band pass filter (equivalent to eFluor™ 450 or Pacific Blue™)
- For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
- If the percentage of dead cells is expected to be less than 5%, then it is recommended to take a small aliquot of cells and heat them at 65°C for 1 minute then immediately place on ice for 1 minute
- After this treatment, the heat-killed cells can be combined 1:1 with live cells and then stained with the Fixable Viability Dye
- Fixable Viability Dye eFluor™ 450 is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO
- It should be protected from light and moisture
- Store at -70°C with dessicant, it may be freeze-thawed up to 20 times
- Allow vial to equilibrate to room temperature before opening

Specifications
Specifications
Color | Blue |
Detection Method | Fluorescence |
For Use With (Application) | Viability Assay |
For Use With (Equipment) | Flow Cytometer |
Product Type | Dye |
Excitation Wavelength Range | 405/450 |
Product Line | eBioscience |
Quantity | 500 Tests |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction