Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FK506 Antibody (FK1), Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FK506 |
---|---|
Clone | FK1 |
Concentration | 1 mg/mL |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017498
![]() |
Novus Biologicals
NB100625710.025MG |
0.025 mg |
Each for $181.90
|
|
|||||
NB10062571
![]() |
Novus Biologicals
NB10062571 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
FK506 Monoclonal specifically detects FK506 in Non-species specific samples. It is validated for ELISA.Specifications
FK506 | |
1 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Non-species specific | |
IgM | |
IgM purified | |
NB100-62571 detects FK-506, a macrolide with similar but more potent immunosuppressant activity than cyclosporine. It is a 23-membered lactone produced by the bacteria Streptomyces tsukubaensis. FK-506 inhibits signal transduction and interleukin-2 transcription in CD4+ T helper lymphocytes, and is mainly used after allogenic organ transplantation to reduce organ rejection. Side effects of FK-506 treatment include nephrotoxicity, neurotoxicity, diabetes mellitus and hypertension. |
FK1 | |
ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
FK-506-BSA conjugate | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title