Learn More
Thermo Scientific™ Fluid Transfer Assembly, Adaptor Set
GREENER_CHOICE
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
$77.00 - $230.00
Specifications
Product Type Adapter Set
View More Specs
Products 9
Catalog Number Mfr. No. Connector Types Tubing Size Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types Tubing Size Price Quantity & Availability  
SH3112101
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31121.01
Luer to MPC TPE 374, 1 m (3.28 ft)
Each for $230.00
Only null left
Add to Cart
SH3112201
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31122.01
Luer to MPC TPE 374, 2 m (6.6 ft)
Each for $77.00
Only null left
Add to Cart
SH3112301
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31123.01
MPX to MPC TPE 374, 2 m (78.7 in)
Each for $81.00
Only null left
Add to Cart
SH3112401
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31124.01
MPX to MPC TPE 374, 2 m (6.6 ft)
Each for $87.00
Only null left
Add to Cart
SH3112501
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31125.01
MPX to MPC C-Flex 374, 2 m (78.7 in)
Each for $81.00
Only null left
Add to Cart
SH3112701
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31127.01
MPX/MPC TPE 374, 2 m (78.7 in)
Each for $83.00
Only null left
Add to Cart
SH3112801
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31128.01
MPX/MPC TPE 374, 2 m (6.6 ft)
Each for $82.00
Only null left
Add to Cart
SH3112901
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31129.01
MPC/MPX TPE 374, 2 m (78.7 in)
Each for $82.00
Only null left
Add to Cart
SH3113001
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31130.01
MPC/MPX TPE 374, 2 m (6.6 ft)
Each for $81.00
Only null left
Add to Cart
Description
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
Adapter Set
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.