Learn More
Eisco™ Fluorite Mineral Specimen
SCI_ED
Catalog No. p-7076818 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Catalog No. S24685
Supplier: Eisco™ ESNG0014
Only null left
Add to Cart
Add to Cart
Fluorite Mineral Specimen is used in classroom identification lab exercises and for study of uncommon 4-plane cleavage pattern.
  • For use in lessons on Bowen's reaction series
  • Has Mohs scale hardness of 4
  • For lessons about hardness and cleavage, as well as the fickle method of identifying minerals by color
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Mineral, Fluorite
For Use With (Application) Study of uncommon 4-plane cleavage pattern
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.