Learn More
Description
- Increased retention for dipolar, unsaturated, or conjugated solutes
- Enhanced selectivity when used with methanolic mobile phase
- Ideal for increasing sensitivity and selectivity in LC-MS analyses
- Heightened selectivity and retention for compounds that are hard to resolve or elute early on C18 and other phenyl chemistries
- Limits ionization suppression and allows simple, MS-friendly mobile phases
- Restek’s most popular LC stationary phase
Specifications
Specifications
| Diameter (Metric) | 2.1 mm |
| For Use With | Bioanalytical Testing Applications |
| Length (Metric) | 100 mm |
| Ligand Type | Biphenyl |
| Max. Pressure | 830 bar (12,000 psi) |
| Particle Size | 1.8 μm |
| pH | 2 to 8 |
| Pore Size | 100 Å |
| Surface Area | 300 m2/g |
| Temperature | 80°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
