Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Force Biphenyl LC Columns, Particle Size: 5 μm
SDP

Catalog No. 06611559 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
06-611-559 150 mm 2.1 mm
06-611-557 50 mm 2.1 mm
06-611-558 50 mm 3 mm
06-611-554 100 mm 2.1 mm
06-611-556 100 mm 3 mm
06-611-555 100 mm 4.6 mm
06-611-561 150 mm 3 mm
06-611-560 150 mm 4.6 mm
06-611-562 250 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 06-611-559 Supplier Restek™ Supplier No. 9629562
Add to Cart
Add to Cart

Ideal for bioanalytical testing applications such as drug and metabolite analyses

  • Increased retention for dipolar, unsaturated, or conjugated solutes
  • Enhanced selectivity when used with methanolic mobile phase
  • Ideal for increasing sensitivity and selectivity in LC-MS analyses
  • Heightened selectivity and retention for compounds that are hard to resolve or elute early on C18 and other phenyl chemistries
  • Limits ionization suppression and allows simple, MS-friendly mobile phases
  • Restek’s most popular LC stationary phase

Specifications

Diameter (Metric) 2.1 mm
For Use With Bioanalytical Testing Applications
Length (Metric) 150 mm
Ligand Type Biphenyl
Max. Pressure 400 bar (5,800 psi)
Particle Size 5 μm
pH 2 to 8
Pore Size 100 Å
Surface Area 300 m2/g
Temperature 80°C
USP Type L11
Stationary Phase Phenyl
Packing Material Fully Porous Silica
Carbon Load 15%
Endcapped Yes
Product Type HPLC/UHPLC Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.