Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO3, p Ser253 Antibody, Novus Biologicals™
SDP

Catalog No. NB017839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017839 0.025 mL
NB10081929 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017839 Supplier Novus Biologicals Supplier No. NB100819290.025ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXO3 Polyclonal specifically detects FOXO3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FOXO3 (p Ser253)
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100
Formulation phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Gene Accession No. O43524
Gene Alias AF6q21, AF6q21 protein, DKFZp781A0677, FKHRL1MGC31925, FKHRL1P2, forkhead box O3, forkhead box protein O3, forkhead homolog (rhabdomyosarcoma) like 1, Forkhead in rhabdomyosarcoma-like 1, forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1, FOXO2, FOXO3Aforkhead box O3A, MGC12739
Gene Symbols FOXO3
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human FKHRL1 around the phosphorylation site of serine 253 (A-V-SP-M-D).
Molecular Weight of Antigen 97 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2309
Test Specificity FKHRL1 (specific to Phospho-Ser253) detects endogenous levels of FKHRL1 only when phosphorylated at serine 253.
Target Species Human, Rat, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.