Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXS1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB031959 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB031959 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB031959 Supplier Novus Biologicals Supplier No. NBP2840310.1ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXS1 Polyclonal specifically detects FOXS1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen FOXS1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias FKHL18, forkhead (Drosophila)-like 18, forkhead box protein S1, forkhead box S1, Forkhead-like 18 protein, forkhead-related activator 10, Forkhead-related transcription factor 10, forkhead-related transcription factor FREAC-10, FREAC-10, FREAC10forkhead-like 18 (Drosophila), MGC4544
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide corresponding to a region of FOXS1
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2307
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.