Learn More
Description
Specifications
Specifications
| Antigen | FXR/NR1H4 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14RXR-interacting protein 14 |
| Gene Symbols | NR1H4 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to the C-terminus of the human FXR protein. [UniProtNo. Q96RI1] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
