Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G2E3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | G2E3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP155281
![]() |
Novus Biologicals
NBP155281 |
100 μL |
Each for $487.50
|
|
|||||
NB15739520
![]() |
Novus Biologicals
NBP15528120UL |
20 μL | N/A | N/A | N/A | ||||
Description
G2E3 Polyclonal specifically detects G2E3 in Human samples. It is validated for Western Blot.Specifications
G2E3 | |
Polyclonal | |
Rabbit | |
Q7L622 | |
55632 | |
Synthetic peptides corresponding to KIAA1333(KIAA1333) The peptide sequence was selected from the N terminal of KIAA1333. Peptide sequence IWQRGKEEEGVYGFLIEDIRKEVNRASKLKCCVCKKNGASIGCVAPRCKR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 6.3.2, EC 6.3.2.-, FLJ20333, G2/M phase-specific E3 ubiquitin-protein ligase, G2/M-phase specific E3 ubiquitin ligase, G2/M-phase specific E3 ubiquitin protein ligase, KIAA1333PHD finger protein 7B, PHF7B | |
G2E3 | |
IgG | |
80 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title