Learn More
Eisco™ Gabbro Specimen
SCI_ED
Catalog No. p-7076849 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
Gabbro forms when iron-rich magma too heavy to reach surface cools slowly over time.
  • Plutonic mafic rock forms from iron-rich magma chambers deep in earth's crust
  • For use in comparing densities between rock samples
  • Example of phaneritic mafic rock
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Igneous Rock, Gabbro
For Use With (Application) Ideal for comparing densities between rock samples
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.