Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAD1/GAD67, Mouse anti-Human, Clone: GAD1/2563, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | GAD1/GAD67 |
---|---|
Clone | GAD1/2563 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Protein Array |
Applications | Western Blot, Flow Cytometry, ELISA, Peptide Array |
Description
GAD1/GAD67 Monoclonal specifically detects GAD1/GAD67 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Protein Array.Specifications
GAD1/GAD67 | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Peptide Array | |
Unconjugated | |
Mouse | |
Diabetes Research | |
10 mM PBS with 0.05% BSA | |
67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
GAD1 | |
IgG2b κ | |
Protein A or G purified | |
This monoclonal antibody recognizes a protein of 67kDa, which is identified as glutamic acid decarboxylase 1 (GAD1/GAD67). |
GAD1/2563 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q99259, Q99259 | |
2571 | |
Recombinant human GAD1/GAD67 protein fragment (around aa 72-135) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title