Learn More
Description
Specifications
Specifications
| Antigen | Galanin R1/GALR1 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 613207 |
| Conjugate | PE |
| Dilution | Flow Cytometry, CyTOF-ready |
| Formulation | PBS |
| Gene Alias | galanin receptor 1, galanin receptor type 1, GALNRGALNR1GAL1-R, GALR-1 |
| Host Species | Mouse |
| Immunogen | NS0 mouse myeloma cell line transfected with human Galanin R1/GALR1, Accession # P47211 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
