Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH Antibody, Novus Biologicals™
SDP

Catalog No. p-5065581 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300326 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300326 Supplier Novus Biologicals Supplier No. NB300326
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

GAPDH Polyclonal specifically detects GAPDH in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunoprecipitation (Negative).

Specifications

Antigen GAPDH
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000, Simple Western 1:25, Immunohistochemistry, Immunoprecipitation, Immunoprecipitation (Negative)
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. P16858
Gene Alias aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Gene Symbols GAPDH
Host Species Rabbit
Immunogen This GAPDH antibody was developed againsted an epitope between residue 300 and the C-terminus of mouse GAPDH using the numbering given in entry NP_001001303.1 (GeneID 407972).
Molecular Weight of Antigen 36 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 2597
Test Specificity Mouse. In tests of lysate from HeLa cells, reactivity with Human GAPDH was weak.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.