Learn More
Eisco™ Garnet Mineral Specimen
SCI_ED
Catalog No. s27577 Shop All Eisco Products
Click to view available options
Quantity:
1/Ea.
12/Pk.
Garnet forms under intense heat and pressure conditions, making it a common mineral in mountainous terranes.
  • Correlates well to Next Generation Science Standards Evidence while Garnet does come in a variety of colors, this specimen is a deep red color
  • Ideal for teaching students about rounding and sorting, as well as for identification & classification exercises
  • Great for lessons in metamorphic facies & silicate minerals.
Specifications
Product Type Mineral
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.