Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva SDS-PAGE Clean-up Kit
Shop All Cytiva Products
Click to view available options
Quantity:
1 Kit
Description
Includes
Precipitant,15mL; Co-precipitant, 15mL; Wash buffer, 50mL; Wash additive, 250μL; Buffer I, 2mL; Buffer II, 500μL; Sample buffer, 2.5mL
- Quantitatively precipitates proteins while leaving interfering substances in solution
- Delivers quantitative yield in less than two hours
Specifications
Specifications
| Boiling Point | 219° to 220°C |
| Color | Colorless |
| Product Type | SDS PAGE Clean-up Kit |
| Form | Liquid |
| Kit Contents | 15mL Precipitant, 15mL coprecipitant, 50mL wash Buffer, 250μL wash additive, 2mL Buffer I, 500μL Buffer II, 2.5mL sample Buffer, quick reference card, instructions |
| Melting Temperature | 168.5°C |
| Molecular Weight (g/mol) | 121.13 |
| Quantity | 1 Kit |
| Storage Requirements | Room Temperature |
| Sufficient For | 50 samples of less than 100μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction