Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP, Mouse anti-All, Clone: 2B6, Millipore Sigma™
Mouse Monoclonal Antibody
$200.50 - $504.50
Specifications
Antigen | GFP |
---|---|
Clone | 2B6 |
Dilution | Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected GFP in HEK293T cells stabely transfected with GFP-tagged Aquaporin 4. |
Applications | Immunocytochemistry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC168925
|
MilliporeSigma
MABC168925UG |
25 μg |
Each for $200.50
|
|
|||||
MABC1689100
|
MilliporeSigma
MABC1689100UG |
100 μg |
Each for $504.50
|
|
|||||
Description
Anti-GFP, clone 2B6, Cat. No. MABC1689, is a mouse monoclonal antibody that detects GFP-conjugated to most proteins and has been tested for use in Immunocytochemistry and Western Blotting.
Green fluorescent protein (UniProt: P42212) is encoded by the GFP gene in Aequorea victoria (Jellyfish). Green fluorescence protein (GFP) is a 27 kDa protein from jellyfish that serves as an energy-transfer acceptor. It transduces the blue chemiluminescence of the protein aequorin into green fluorescent light by energy transfer and it fluoresces in vivo upon receiving energy from the Ca2+-activated photoprotein aequorin. GFP has an absorption max at 395 nm and the emission peaks at 507 to 510 nm. However, the exact value of emission max depends on the environment of the chromophore. GFP is an important tool in cell biology research that displays stability under fixation conditions and is suitable for a variety of applications, such as tracing cell lineage tracer, reporter of gene expression, or exploring protein-protein interactions.Specifications
GFP | |
Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected GFP in HEK293T cells stabely transfected with GFP-tagged Aquaporin 4. | |
Monoclonal | |
Purified | |
RUO | |
All species | |
Green fluorescent protein | |
His-tagged recombinant jellyfish Green fluorescent protein. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
2B6 | |
Immunocytochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Secondary & Control Antibodies | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
GFP | |
IgG1 κ | |
Protein G purified | |
Clone 2B6 is a mouse monoclonal antibody that detects GFP-conjugated to most proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title