Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GJD3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen GJD3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15916020
SDP
View Documents
Novus Biologicals
NBP15916020UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP159160
SDP
View Documents
Novus Biologicals
NBP159160
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

GJD3 Polyclonal specifically detects GJD3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

GJD3
Polyclonal
Purified
RUO
Connexin-31.9, Cx30.2, Cx31.9, CX31.9connexin-31.9, Gap junction alpha-11 protein, Gap junction chi-1 protein, gap junction protein, chi 1, 31.9kDa, gap junction protein, delta 3, 31.9kDa, GJA11connexin 31.9, GJC1gap junction delta-3 protein
GJD3
IgG
Protein A purified
Western Blot
Unconjugated
Rabbit
Q6ZUW6
125111
Synthetic peptides corresponding to GJC1 (gap junction protein, chi 1, 31.9kDa) The peptide sequence was selected from the N terminal of GJC1. Peptide sequence IFRILVLATVGGAVFEDEQEEFVCNTLQPGCRQTCYDRAFPVSHYRFWLF.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.