Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI-1 Antibody (CL12191), Novus Biologicals™
SDP

Catalog No. NB236182 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB236182 100 μg
NB236828 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB236182 Supplier Novus Biologicals Supplier No. NBP321210100UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLI-1 Monoclonal antibody specifically detects GLI-1 in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen GLI-1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone CL12191
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI
Host Species Mouse
Immunogen This antibody was developed against a Synthetic Peptide of P08151. Actual immunogen sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2735
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.