Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLUT12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00
Specifications
Antigen | GLUT12 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16002320
![]() |
Novus Biologicals
NBP16002320UL |
20 μL |
Each for $158.00
|
|
|||||
NBP160023
![]() |
Novus Biologicals
NBP160023 |
100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
GLUT12 Polyclonal specifically detects GLUT12 in Human samples. It is validated for Western Blot.Specifications
GLUT12 | |
Polyclonal | |
Rabbit | |
Q8TD20 | |
154091 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
GLUT-12, GLUT12GLUT8Glucose transporter type 12, solute carrier family 2 (facilitated glucose transporter), member 12, solute carrier family 2, facilitated glucose transporter member 12 | |
SLC2A12 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title