Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GNF 5837

Click to view available options
Quantity:
10 mg
50 mg
Description
Potent pan-Trk inhibitor. Displays antiproliferative effects in cellular Ba/F3 assays (IC50 values are 7, 9 and 11 nM for cells containing the fusion proteins Tel-TrkC, Tel-TrkB and Tel-TrkA, respectively). Exhibits selectivity for Trk receptors over a range of kinases, with some activity at PDGFR and c-Kit (IC50 values are 0.87 and 0.91 μM respectively). Orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 535.49 |
| Purity | >98% |
| Molecular Weight (g/mol) | 535.50 |
| Chemical Name or Material | GNF 5837 |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction