Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPCR/G-protein complex-stabilizing scFv Recombinant Mouse Monoclonal Antibody (scFv16)

Mouse Recombinant Monoclonal Antibody

$378.65 - $2035.00

Specifications

Antigen GPCR/G-protein complex-stabilizing scFv
Clone scFv16
Concentration 2.0 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications Not Available
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
70-397-6
Encompass_Preferred
View Documents
Invitrogen™
703976
100 μg
Each for $378.65

Save 25%

Reg : $504.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart
 
7039761MG
Encompass_Preferred
View Documents
Invitrogen™
7039761MG
1 mg
Each for $2,035.00
Only null left
Add to Cart
 
Description

Description

The scFv antibody stabilizes GPCR/G-protein complexes for cryo-EM of active structures. It recognizes an interface between G alpha and G beta/gamma subunits, and confers resistance to nucleotide exchange-triggered dissociation of the heterotrimeric G-protein. In addition to members of the Gi/o family, this antibody fragment can also be applied to chimeric variants and stabilize other G-protein subtypes including Gs complexes.

Specifications

Specifications

GPCR/G-protein complex-stabilizing scFv
2.0 mg/mL
Not Available
Unconjugated
Mouse
Human
Rhodopsin/Gi1.
Antibody
scFv16
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
RUO
25mM HEPES with 10% glycerol and no preservative; pH 7.5
Primary
Affinity Chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.