Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan Scientific and Medical™ Graphite Vespel™ Ferrules for Shimadzu
SDP

Catalog No. 0726561
Click to view available options
Diameter (Metric) Inside:
0.10 to 0.25 mm
0.32 mm

Provides leak-free system connections. Trajan Scientific and Medical Graphite Vespel™ Ferrules for Shimadzu leads to improved chromatography for GC systems.

  • Composite of graphite and Vespel
  • Mechanically robust, hard material, long lifetime
  • Forms a strong grip with the capillary column

Specifications

Material 15% Graphite, 85% Vespel
For Use With (Equipment) QP5000-II and QP5050
Type Graphite Vespel Ferrule for 0.1-0.25mm ID Columns for Shimadzu MS
Diameter (Metric) Inside 0.10 to 0.25 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.