Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Graphsphere™ 2016 Glass Thermal Desorption Tubes
Shop All MilliporeSigma Supelco Products
Click to view available options
Description:
Sealed with Brass Endcaps
Sealed with TDS3 Storage Container
Specifications
Specifications
| Diameter (English) Outer | 1/4 in. |
| Material | Glass |
| Packaging Quantity | 1 Ea. |
| Overall Length (English) | 3.5 in. |
| Matrix | Carboxen™ 1016 Carbon Molecular Sieve (CMS) |
| For Use With (Application) | An Alternative to Tenax™ TA |
| Description | Sealed with Brass Endcaps |
| For Use With (Equipment) | PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments |
| Type | Thermal Desorption Tube |
Graphsphere is a trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction