Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRASP55 Antibody (CL2522), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | GRASP55 |
---|---|
Clone | CL2522 |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000-1:10000 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
GRASP55 Monoclonal specifically detects GRASP55 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GRASP55 | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q9H8Y8 | |
26003 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PTRPFEEGKKISLPGQMAGTPITPLKDGFTEVQLSSVNPPSLSPPGTTGIEQSLTGLSISSTPPAVSSVLSTGV | |
Primary | |
Protein A purified |
CL2522 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Golgi Apparatus Markers | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Golgi phosphoprotein 6, golgi reassembly stacking protein 2, 55 kDa, golgi reassembly stacking protein 2, 55kDa, Golgi reassembly-stacking protein of 55 kDa, GOLPH6DKFZp434D156, GRASP55FLJ13139, GRS2Golgi reassembly-stacking protein 2, p59 | |
GORASP2 | |
IgG1 | |
Protein A purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title