Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | GRK4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15531320
![]() |
Novus Biologicals
NBP15531320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155313
![]() |
Novus Biologicals
NBP155313 |
100 μL |
Each for $487.50
|
|
|||||
Description
GRK4 Polyclonal specifically detects GRK4 in Human samples. It is validated for Western Blot.Specifications
GRK4 | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
EC 2.7.11, EC 2.7.11.16, G protein-coupled receptor kinase 2-like (Drosophila), G protein-coupled receptor kinase 4, G protein-coupled receptor kinase GRK4, GPRK2LIT11, GPRK4GRK4a, G-protein coupled receptor kinase 4, ITI1 | |
GRK4 | |
IgG | |
59 kDa |
Western Blot | |
Unconjugated | |
RUO | |
P32298 | |
2868 | |
Synthetic peptides corresponding to GRK4(G protein-coupled receptor kinase 4) The peptide sequence was selected from the middle region of GRK4. Peptide sequence QSRFVVSLAYAYETKDALCLVLTIMNGGDLKFHIYNLGNPGFDEQRAVFY. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title