Learn More
Description
Specifications
Specifications
| Antigen | GroEL |
| Applications | Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 9A1/2 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:10000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500 |
| Formulation | PBS and 50% Glycerol with 0.09% Sodium Azide |
| Gene Alias | Cpn60 chaperonin GroEL, large subunit of GroESL, ECK4137, groEL, JW4103, mopA |
| Host Species | Mouse |
| Immunogen | E. coli GroEL. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
